4-Cyclopentyl-1-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48540-500MG 48540-1G 48540-25G 48540-5G
77327-588EA 298.7 USD
77327-588 77351-186 77386-362 77375-476
4-Cyclopentyl-1-fluoro-2-methylbenzene ≥97%
4-Cyclopentyl-1-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₅F
MW: 178.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902745
CAS Number: 2586127-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR