1-(5-Bromo-2-fluoro-3-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

48533-500MG 48533-1G 48533-25G 48533-5G
77327-586EA 299.73 USD
77327-586 77351-182 77386-360 77375-472
1-(5-Bromo-2-fluoro-3-methylphenyl)ethanol ≥95%
1-(5-Bromo-2-fluoro-3-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141732
CAS Number: 1252572-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR