4-Fluoro-3-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48498-500MG 48498-1G 48498-5G
77336-676EA 391.4 USD
77336-676 77351-176 77375-468
4-Fluoro-3-(methoxymethyl)benzaldehyde ≥97%
4-Fluoro-3-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529980
CAS Number: 1891258-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR