Methyl 2,6-dibromo-4-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48497-500MG 48497-1G 48497-5G 48497-25G
77327-580EA 134.93 USD
77327-580 77344-462 77363-918 77386-356
Methyl 2,6-dibromo-4-isopropylbenzoate ≥97%
Methyl 2,6-dibromo-4-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32902743
CAS Number: 2586126-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR