4-Bromo-2-chloro-3-methylaniline ≥95%

Supplier: AOBChem USA

Warning

48482-500MG 48482-1G 48482-5G 48482-25G
77327-578EA 223.51 USD
77327-578 77351-174 77363-916 77386-354
4-Bromo-2-chloro-3-methylaniline ≥95%
4-Bromo-2-chloro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.494 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30478267
CAS Number: 80026-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR