5-Bromo-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

48446-500MG 48446-1G 48446-5G 48446-25G
77327-576EA 204.97 USD
77327-576 77351-172 77363-914 77386-352
5-Bromo-2-(methylthio)phenol ≥97%
5-Bromo-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₇BrOS
MW: 219.099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999030
CAS Number: 107724-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR