4-(tert-Butyl)-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

48436-500MG 48436-1G 48436-25G 48436-5G
77327-574EA 285.31 USD
77327-574 77351-170 77386-350 77375-466
4-(tert-Butyl)-2-methoxyphenylboronic acid ≥97%
4-(tert-Butyl)-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
MW: 208.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11043447
CAS Number: 1245928-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR