Ethyl 2,6-difluoro-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

48394-500MG 48394-1G 48394-5G 48394-25G
77327-564EA 187.46 USD
77327-564 77351-160 77363-904 77386-340
Ethyl 2,6-difluoro-4-(methylthio)benzoate ≥97%
Ethyl 2,6-difluoro-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34529977
CAS Number: 2734777-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR