Ethyl 4-methyl-3-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

48392-500MG 48392-1G 48392-5G
77337-406EA 414.06 USD
77337-406 77351-156 77375-462
Ethyl 4-methyl-3-(methylthio)benzoate ≥95%
Ethyl 4-methyl-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489179
CAS Number: 1824579-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR