Methyl 4-methyl-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

48391-500MG 48391-1G 48391-5G
77337-404EA 414.06 USD
77337-404 77351-154 77375-460
Methyl 4-methyl-3-(methylthio)benzoate ≥97%
Methyl 4-methyl-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
MW: 196.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489130
CAS Number: 1824661-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR