(2-(Benzyloxy)-4-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48382-500MG 48382-1G 48382-5G
77336-186EA 380.07 USD
77336-186 77351-148 77375-458
(2-(Benzyloxy)-4-fluorophenyl)methanol ≥97%
(2-(Benzyloxy)-4-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701706
CAS Number: 351445-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR