Ethyl 2-(benzyloxy)-4-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

48379-500MG 48379-1G 48379-5G 48379-25G
77327-554EA 281.19 USD
77327-554 77351-144 77363-896 77386-330
Ethyl 2-(benzyloxy)-4-fluorobenzoate ≥97%
Ethyl 2-(benzyloxy)-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₃
MW: 274.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529975
CAS Number: 2734774-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR