Ethyl 2-ethoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

48375-500MG 48375-1G 48375-5G 48375-25G
77327-548EA 225.57 USD
77327-548 77351-138 77363-890 77386-324
Ethyl 2-ethoxy-5-methylbenzoate ≥97%
Ethyl 2-ethoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28967290
CAS Number: 854645-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR