Ethyl 2-bromo-5-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48371-500MG 48371-1G 48371-5G 48371-25G
77327-544EA 120.51 USD
77327-544 77343-724 77363-886 77375-456
Ethyl 2-bromo-5-hydroxybenzoate ≥97%
Ethyl 2-bromo-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566801
CAS Number: 102297-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR