Ethyl 4-bromo-5-methylthiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

48369-500MG 48369-1G 48369-5G 48369-25G
77327-542EA 281.19 USD
77327-542 77351-134 77363-884 77386-320
Ethyl 4-bromo-5-methylthiophene-2-carboxylate ≥97%
Ethyl 4-bromo-5-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉BrO₂S
MW: 249.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25368844
CAS Number: 1256286-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR