(4-Methoxy-[1,1'-biphenyl]-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

48368-500MG 48368-1G 48368-5G 48368-25G
77327-540EA 225.57 USD
77327-540 77351-132 77363-882 77386-318
(4-Methoxy-[1,1'-biphenyl]-2-yl)methanol ≥97%
(4-Methoxy-[1,1'-biphenyl]-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203076
CAS Number: 52008-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR