2,6-Difluoro-3-(methylthio)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

48359-500MG 48359-1G 48359-5G 48359-25G
77327-538EA 145.23 USD
77327-538 77345-076 77363-880 77386-316
2,6-Difluoro-3-(methylthio)benzonitrile ≥95%
2,6-Difluoro-3-(methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂NS
Storage Temperature: Ambient
MDL Number: MFCD28765124
CAS Number: 1520381-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR