2-Fluoro-3-formyl-6-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

48353-500MG 48353-1G 48353-5G 48353-25G
77327-534EA 265.74 USD
77327-534 77351-130 77363-876 77386-312
2-Fluoro-3-formyl-6-methoxybenzonitrile ≥97%
2-Fluoro-3-formyl-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD34178541
CAS Number: 1520916-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR