Methyl 2-chloro-4,5-dimethoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

48332-500MG 48332-1G 48332-5G 48332-25G
77327-530EA 181.28 USD
77327-530 77346-882 77363-872 77386-308
Methyl 2-chloro-4,5-dimethoxybenzoate ≥95%
Methyl 2-chloro-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD15530341
CAS Number: 30714-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR