2-Chloro-4,5-dimethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

48330-500MG 48330-1G 48330-5G
77336-062EA 375.95 USD
77336-062 77351-128 77375-454
2-Chloro-4,5-dimethoxyphenol ≥97%
2-Chloro-4,5-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD24721193
CAS Number: 18093-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR