(4-Bromo-2,3-dichlorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48297-500MG 48297-1G 48297-5G
77337-272EA 410.97 USD
77337-272 77351-126 77375-452
(4-Bromo-2,3-dichlorophenyl)methanol ≥97%
(4-Bromo-2,3-dichlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737837
CAS Number: 1807172-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR