(2,6-difluoro-3-propoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

48282-500MG 48282-1G 48282-5G
77337-908EA 451.14 USD
77337-908 77351-124 77375-450
(2,6-difluoro-3-propoxyphenyl)methanol ≥95%
(2,6-difluoro-3-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403877
CAS Number: 2635937-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR