2,6-Difluoro-3-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48281-500MG 48281-1G 48281-5G 48281-25G
77327-526EA 158.62 USD
77327-526 77345-736 77363-868 77386-304
2,6-Difluoro-3-propoxybenzaldehyde ≥97%
2,6-Difluoro-3-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707322
CAS Number: 2484888-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR