Methyl 2,6-difluoro-3-propoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

48279-500MG 48279-1G 48279-5G 48279-25G
77327-522EA 226.6 USD
77327-522 77351-120 77363-864 77386-300
Methyl 2,6-difluoro-3-propoxybenzoate ≥95%
Methyl 2,6-difluoro-3-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529970
CAS Number: 2734776-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR