1-Chloro-5-fluoro-2-iodo-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

48276-500MG 48276-1G 48276-5G
77335-744EA 364.62 USD
77335-744 77351-118 77375-446
1-Chloro-5-fluoro-2-iodo-3-(trifluoromethyl)benzene ≥97%
1-Chloro-5-fluoro-2-iodo-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂ClF₄I
MW: 324.442 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529968
CAS Number: 2386335-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR