1,2-Dichloro-5-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

48274-500MG 48274-1G 48274-5G
77337-556EA 421.27 USD
77337-556 77351-116 77375-444
1,2-Dichloro-5-fluoro-3-(trifluoromethyl)benzene ≥97%
1,2-Dichloro-5-fluoro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₄
Storage Temperature: Ambient
MDL Number: MFCD28786013
CAS Number: 1804886-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR