5-(Benzyloxy)-6-methylpicolinic acid ≥97%

Supplier: AOBChem USA

Warning

48255-500MG 48255-1G 48255-5G 48255-25G
77327-514EA 281.19 USD
77327-514 77351-110 77363-858 77386-296
5-(Benzyloxy)-6-methylpicolinic acid ≥97%
5-(Benzyloxy)-6-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MW: 243.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258403
CAS Number: 1256808-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR