5-(Benzyloxy)-6-methylpicolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

48253-500MG 48253-1G 48253-5G 48253-25G
77327-512EA 188.49 USD
77327-512 77351-108 77363-856 77386-294
5-(Benzyloxy)-6-methylpicolinaldehyde ≥97%
5-(Benzyloxy)-6-methylpicolinaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258402
CAS Number: 59781-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR