(2-Ethoxy-5-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48251-500MG 48251-1G 48251-5G 48251-25G
77327-510EA 251.32 USD
77327-510 77351-106 77363-854 77386-292
(2-Ethoxy-5-fluorophenyl)methanol ≥97%
(2-Ethoxy-5-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11639967
CAS Number: 1096330-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR