2-Fluoro-5-methyl-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48244-500MG 48244-1G 48244-5G
77336-148EA 379.04 USD
77336-148 77351-098 77375-436
2-Fluoro-5-methyl-3-(methylthio)benzaldehyde ≥97%
2-Fluoro-5-methyl-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FOS
MW: 184.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403874
CAS Number: 2092104-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR