benzyl 5-bromo-6-chloronicotinate ≥97%

Supplier: AOBChem USA

Warning

48236-500MG 48236-1G 48236-5G 48236-25G
77327-502EA 100.94 USD
77327-502 77342-916 77363-846 77375-434
benzyl 5-bromo-6-chloronicotinate ≥97%
benzyl 5-bromo-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₁₃H₉BrClNO₂
MW: 326.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529967
CAS Number: 1852408-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR