5-Bromo-6-chloro-N-cyclohexylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

48235-500MG 48235-1G 48235-5G 48235-25G
77327-500EA 211.15 USD
77327-500 77351-096 77363-844 77386-286
5-Bromo-6-chloro-N-cyclohexylnicotinamide ≥97%
5-Bromo-6-chloro-N-cyclohexylnicotinamide

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrClN₂O
MW: 317.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756416
CAS Number: 2024991-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR