5-Bromo-6-chloro-N,N-dimethylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

48234-500MG 48234-1G 48234-5G 48234-25G
77327-498EA 211.15 USD
77327-498 77351-094 77363-842 77386-284
5-Bromo-6-chloro-N,N-dimethylnicotinamide ≥97%
5-Bromo-6-chloro-N,N-dimethylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₈BrClN₂O
MW: 263.519 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551404
CAS Number: 1851868-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR