(5-Bromo-6-chloropyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48233-500MG 48233-1G 48233-5G 48233-25G
77327-496EA 211.15 USD
77327-496 77351-092 77363-840 77386-282
(5-Bromo-6-chloropyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%
(5-Bromo-6-chloropyridin-3-yl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂O
MW: 289.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551411
CAS Number: 1856976-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR