(5-Bromo-6-chloropyridin-3-yl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

48232-500MG 48232-1G 48232-5G 48232-25G
77327-494EA 211.15 USD
77327-494 77351-090 77363-838 77386-280
(5-Bromo-6-chloropyridin-3-yl)(piperidin-1-yl)methanone ≥97%
(5-Bromo-6-chloropyridin-3-yl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClN₂O
MW: 303.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551413
CAS Number: 1872711-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR