(5-Bromo-6-chloropyridin-3-yl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

48231-500MG 48231-1G 48231-5G 48231-25G
77327-492EA 211.15 USD
77327-492 77351-088 77363-836 77386-278
(5-Bromo-6-chloropyridin-3-yl)(morpholino)methanone ≥97%
(5-Bromo-6-chloropyridin-3-yl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂O₂
MW: 305.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551414
CAS Number: 1875938-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR