(5-Bromo-6-chloropyridin-3-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

48230-500MG 48230-1G 48230-5G
77327-490EA 334.75 USD
77327-490 77351-086 77375-432
(5-Bromo-6-chloropyridin-3-yl)(thiomorpholino)methanone ≥97%
(5-Bromo-6-chloropyridin-3-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂OS
MW: 321.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756415
CAS Number: 1882727-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR