(5-Bromo-6-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48229-500MG 48229-1G 48229-5G 48229-25G
77327-488EA 221.45 USD
77327-488 77351-084 77363-834 77386-276
(5-Bromo-6-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥95%
(5-Bromo-6-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClN₃O
MW: 318.597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551415
CAS Number: 2025208-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR