5-Bromo-6-chloro-N-cyclopropylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

48228-500MG 48228-1G 48228-5G 48228-25G
77327-486EA 211.15 USD
77327-486 77351-082 77363-832 77386-274
5-Bromo-6-chloro-N-cyclopropylnicotinamide ≥97%
5-Bromo-6-chloro-N-cyclopropylnicotinamide

Precaución: for Research Use Only

Formula: C₉H₈BrClN₂O
MW: 275.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33551406
CAS Number: 1864195-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR