5-Bromo-6-chloro-N-methylnicotinamide ≥97%
Supplier: AOBChem USA
|
Warning
|
48227-500MG
48227-1G
48227-5G
48227-25G
77327-484EA
201.88
USD
77327-484
77351-080
77363-830
77386-272
5-Bromo-6-chloro-N-methylnicotinamide ≥97%
5-Bromo-6-chloro-N-methylnicotinamide
Precaución: for Research Use Only
Formula:
C₇H₆BrClN₂O MW: 249.492 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33551403 CAS Number: 1881579-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...