(2-Methoxy-5-methylpyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

48212-500MG 48212-1G 48212-5G 48212-25G
77327-480EA 243.08 USD
77327-480 77351-076 77363-826 77386-268
(2-Methoxy-5-methylpyridin-3-yl)methanol ≥97%
(2-Methoxy-5-methylpyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD16611529
CAS Number: 351410-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR