(5-Bromo-2-chloropyridin-4-yl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

48206-500MG 48206-1G 48206-5G 48206-25G
77327-474EA 99.91 USD
77327-474 77342-798 77363-820 77375-424
(5-Bromo-2-chloropyridin-4-yl)(morpholino)methanone ≥97%
(5-Bromo-2-chloropyridin-4-yl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂O₂
MW: 305.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707318
CAS Number: 1870258-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR