(5-Bromo-2-chloropyridin-4-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

48205-500MG 48205-1G 48205-5G 48205-25G
77327-472EA 122.57 USD
77327-472 77343-912 77363-818 77375-422
(5-Bromo-2-chloropyridin-4-yl)(thiomorpholino)methanone ≥97%
(5-Bromo-2-chloropyridin-4-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂OS
MW: 321.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707317
CAS Number: 1871486-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR