(5-Bromo-2-chloropyridin-4-yl)(4-methylpiperazin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48204-500MG 48204-1G 48204-5G 48204-25G
77327-470EA 143.17 USD
77327-470 77344-984 77363-816 77386-266
(5-Bromo-2-chloropyridin-4-yl)(4-methylpiperazin-1-yl)methanone ≥95%
(5-Bromo-2-chloropyridin-4-yl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClN₃O
MW: 318.597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707316
CAS Number: 1996879-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR