5-Bromo-2-chloro-N-cyclopropylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

48203-500MG 48203-1G 48203-5G 48203-25G
77327-468EA 97.85 USD
77327-468 77342-660 77363-814 77375-420
5-Bromo-2-chloro-N-cyclopropylisonicotinamide ≥97%
5-Bromo-2-chloro-N-cyclopropylisonicotinamide

Precaución: for Research Use Only

Formula: C₉H₈BrClN₂O
MW: 275.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707315
CAS Number: 1862872-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR