(5-Bromo-2-chloropyridin-4-yl)(pyrrolidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

48201-1G 48201-5G 48201-25G
77341-968EA 109.18 USD
77341-968 77363-812 77375-418
(5-Bromo-2-chloropyridin-4-yl)(pyrrolidin-1-yl)methanone ≥95%
(5-Bromo-2-chloropyridin-4-yl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClN₂O
MW: 289.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707313
CAS Number: 1875545-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR