2,4-Difluoro-1-(1-(4-methoxyphenyl)ethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

48136-500MG 48136-1G 48136-5G
77337-358EA 412 USD
77337-358 77351-066 77375-406
2,4-Difluoro-1-(1-(4-methoxyphenyl)ethyl)benzene ≥95%
2,4-Difluoro-1-(1-(4-methoxyphenyl)ethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄F₂O
Storage Temperature: Ambient
MDL Number: MFCD32707306
CAS Number: 2484888-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR