2,4-Difluoro-1-(1-(p-tolyl)ethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

48135-500MG 48135-1G 48135-5G
77335-502EA 356.38 USD
77335-502 77351-064 77375-404
2,4-Difluoro-1-(1-(p-tolyl)ethyl)benzene ≥95%
2,4-Difluoro-1-(1-(p-tolyl)ethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄F₂
Storage Temperature: Ambient
MDL Number: MFCD32707305
CAS Number: 2484888-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR