4-(1-(2,5-difluorophenyl)ethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48134-500MG 48134-1G 48134-5G
77336-194EA 381.1 USD
77336-194 77351-062 77375-402
4-(1-(2,5-difluorophenyl)ethyl)benzaldehyde ≥95%
4-(1-(2,5-difluorophenyl)ethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD32707304
CAS Number: 2484888-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR