4-(1-(3-bromo-4-methoxyphenyl)ethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48130-500MG 48130-1G 48130-5G
77336-146EA 379.04 USD
77336-146 77351-058 77375-398
4-(1-(3-bromo-4-methoxyphenyl)ethyl)benzaldehyde ≥95%
4-(1-(3-bromo-4-methoxyphenyl)ethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrO₂
MW: 319.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707300
CAS Number: 2484888-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR